Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EIF4G3 SIRNA Assays ›
Gene Symbol:
EIF4G3
Gene Name:
eukaryotic translation initiation factor 4 gamma 3
Gene Aliases:
eIF-4G 3, eIF4G 3, eIF4GII
Chromosome Location:
Chr.1: 20806292 - 21177320 on Build GRCh38
UniGene ID:
Hs.467084
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16519
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EIF4G3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001198801.1
NP_001185730.1
25, 26
4110
NM_001198802.1
NP_001185731.1
25, 26
4020
NM_003760.4
NP_003751.2
22, 23
3636
XM_011542356.2
XP_011540658.1
28, 29
4710
XM_011542361.2
XP_011540663.1
27, 28
4599
XM_011542370.1
XP_011540672.1
28, 29
4713
XM_011542371.1
XP_011540673.1
26, 27
4455
XM_011542374.1
XP_011540676.1
25, 26
4422
XM_011542375.2
XP_011540677.1
25, 26
6010
XM_011542381.2
XP_011540683.1
15, 16
2096
XM_017002672.1
XP_016858161.1
29, 30
6248
XM_017002673.1
XP_016858162.1
27, 28
4692
XM_017002674.1
XP_016858163.1
27, 28
4689
XM_017002675.1
XP_016858164.1
27, 28
4680
XM_017002676.1
XP_016858165.1
26, 27
4659
XM_017002677.1
XP_016858166.1
26, 27
4536
XM_017002678.1
XP_016858167.1
27, 28
4602
XM_017002679.1
XP_016858168.1
27, 28
4599
XM_017002680.1
XP_016858169.1
24, 25
3629
XM_017002681.1
XP_016858170.1
27, 28
4596
XM_017002682.1
XP_016858171.1
26, 27
4578
XM_017002683.1
XP_016858172.1
26, 27
4578
XM_017002684.1
XP_016858173.1
27, 28
6104
XM_017002685.1
XP_016858174.1
26, 27
4569
XM_017002686.1
XP_016858175.1
26, 27
4566
XM_017002687.1
XP_016858176.1
26, 27
4566
XM_017002688.1
XP_016858177.1
23, 24
3595
XM_017002689.1
XP_016858178.1
28, 29
4713
XM_017002690.1
XP_016858179.1
26, 27
4563
XM_017002691.1
XP_016858180.1
25, 26
4548
XM_017002692.1
XP_016858181.1
25, 26
4545
XM_017002693.1
XP_016858182.1
24, 25
4416
XM_017002694.1
XP_016858183.1
26, 27
4488
XM_017002695.1
XP_016858184.1
26, 27
4485
XM_017002696.1
XP_016858185.1
25, 26
4467
XM_017002697.1
XP_016858186.1
25, 26
4464
XM_017002698.1
XP_016858187.1
25, 26
4455
XM_017002699.1
XP_016858188.1
27, 28
4602
XM_017002700.1
XP_016858189.1
24, 25
4434
XM_017002701.1
XP_016858190.1
24, 25
4431
XM_017002702.1
XP_016858191.1
24, 25
4365
XM_017002703.1
XP_016858192.1
22, 23
3526
XM_017002704.1
XP_016858193.1
26, 27
4578
XM_017002705.1
XP_016858194.1
27, 28
3873
XM_017002706.1
XP_016858195.1
26, 27
3762
XM_017002707.1
XP_016858196.1
24, 25
3705
XM_017002708.1
XP_016858197.1
23, 24
3594
XM_017002709.1
XP_016858198.1
25, 26
4457
GenBank mRNA
AB209119.1
-
25, 26
3979
AF012072.2
-
22, 23
3636
AK294883.1
-
16, 17
2883
AK302873.1
-
13, 14
1877
BC094683.1
-
20, 21
2643
6188273
BC136643.1
-
22, 23
3559
9020633
Z34918.1
-
6, 7
722
Back To Top
More Information
Panther Classification:
Molecular Function -
translation factor
translation initiation factor
translational protein
Gene Ontology Categories:
Function(s)
Process(es)
translational initiation
regulation of translational initiation
viral process
RNA cap binding
translation initiation factor activity
translation factor activity, RNA binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0