Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1565430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257538.2 | XP_006257600.1 | 468 | |||
XM_008773538.1 | XP_008771760.1 | 468 |
Gene Symbol: LOC100364877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752159.3 | XP_003752207.1 | 1 | 323 |
Gene Symbol: LOC103694516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773552.1 | XP_008771774.1 | 1 | 323 |
Gene Symbol: LOC103694587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773722.1 | XP_008771944.1 | 1 | 323 |
Gene Symbol: LOC691030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773536.1 | XP_008771758.1 | 1 | 461 |
Gene Symbol: RGD1561558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_228669.5 | XP_228669.3 | 323 |