Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3h11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047902.1 | NP_001041367.1 | 9 | 1105 | ||
XM_008769482.1 | XP_008767704.1 | 7 | 882 | |||
XM_008769483.1 | XP_008767705.1 | 7 | 879 | |||
XM_008769484.1 | XP_008767706.1 | 7 | 855 | |||
GenBank mRNA | BC111404.1 | - | 2 | 118 | ||
DQ480752.1 | - | 9 | 1105 |