Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251004.2 | XP_006251066.1 | 2 | 654 | ||
XM_006251005.2 | XP_006251067.1 | 2 | 654 | |||
XM_341215.7 | XP_341216.4 | 2 | 654 |