Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251004.2 | XP_006251066.1 | 17 | 3036 | ||
XM_006251005.2 | XP_006251067.1 | 16 | 2871 | |||
XM_006251006.2 | XP_006251068.1 | 16 | 2430 | |||
XM_006251007.1 | XP_006251069.1 | 16 | 2454 | |||
XM_341215.7 | XP_341216.4 | 15 | 2751 |