Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ASMT SIRNA Assays ›
          Gene Symbol:
          ASMT ARRDC3-AS1 ELMOD2 FNIP2 LPP TACC2
          Gene Name:
          acetylserotonin O-methyltransferase
          ARRDC3 antisense RNA 1
          ELMO domain containing 2
          folliculin interacting protein 2
          LIM domain containing preferred translocation partner in lipoma
          transforming acidic coiled-coil containing protein 2
          Gene Aliases:
          ASMTY, HIOMT, HIOMTY
          -
          9830169G11Rik
          FNIPL, MAPO1
          -
          AZU-1, ECTACC
          Chromosome Location:
          Chr.Y: 1595455 - 1643081 on Build GRCh38
          Chromosome Location:
          Chr.5: 91380347 - 91420715 on Build GRCh38
          Chromosome Location:
          Chr.4: 140524146 - 140553770 on Build GRCh38
          Chromosome Location:
          Chr.4: 158768995 - 158908050 on Build GRCh38
          Chromosome Location:
          Chr.3: 188153050 - 188890671 on Build GRCh38
          Chromosome Location:
          Chr.10: 121989174 - 122254545 on Build GRCh38
          UniGene ID:
          Hs.522572
          Hs.116364
          Hs.450105
          Hs.652441
          Hs.720220
          Hs.501252
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          -
          -
          -
          -
          siRNA ID s1672

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ASMT
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001171038.1 NP_001164509.1 6 766
          NM_004043.2 NP_004034.2 7 759
          GenBank mRNA AK314922.1 - 6 767
          M83779.1 - 6 683
          U11090.1 - 7 759
          U11091.1 - 6 717
          Gene Symbol: ARRDC3-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027435.1 3 2209
          GenBank mRNA BC043415.2 - 3 2198
          Gene Symbol: ELMOD2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq XM_011531818.2 XP_011530120.1 9 3463
          GenBank mRNA AK021525.1 - 1 1532
          Gene Symbol: FNIP2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC007861.1 - 3 1379
          Gene Symbol: LPP
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BX474180.1 - 3 322
          Gene Symbol: TACC2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK094848.1 - 2 1690

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          methyltransferase scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          translation
          melatonin biosynthetic process
          methylation
          indolalkylamine biosynthetic process
          positive regulation of GTPase activity
          regulation of defense response to virus
          defense response to virus
          O-methyltransferase activity
          S-methyltransferase activity
          acetylserotonin O-methyltransferase activity
          identical protein binding
          protein homodimerization activity
          GTPase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-4sktr:80/100.66.76.202:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0