Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006230081.2 | XP_006230143.1 | 3 | 881 | ||
XM_006230082.2 | XP_006230144.1 | 3 | 881 | |||
XM_006230084.2 | XP_006230146.1 | 3 | 660 | |||
XM_008759769.1 | XP_008757991.1 | 3 | 881 | |||
GenBank mRNA | BC168986.1 | - | 2 | 288 | ||
FQ219437.1 | - | 2 | 184 |