Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bccip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108505.2 | NP_001101975.2 | 7 | 897 | ||
GenBank mRNA | FQ227185.1 | - | 7 | 888 | ||
FQ228402.1 | - | 7 | 895 |