Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ebf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108506.1 | NP_001101976.1 | 15 | 1930 | ||
XM_006230462.2 | XP_006230524.1 | 14 | 2083 | |||
XM_006230463.2 | XP_006230525.1 | 14 | 2082 | |||
XM_006230464.2 | XP_006230526.1 | 14 | 2054 | |||
XM_006230465.2 | XP_006230527.1 | 14 | 2053 | |||
XM_008759997.1 | XP_008758219.1 | 14 | 2084 | |||
XM_008759998.1 | XP_008758220.1 | 14 | 2056 | |||
XM_008759999.1 | XP_008758221.1 | 14 | 2083 |