Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013194.1 | NP_001013212.1 | 2 | 827 | ||
XM_006230820.1 | XP_006230882.1 | 3 | 1940 | |||
XM_006230821.1 | XP_006230883.1 | 2 | 2159 | |||
XM_006230822.1 | XP_006230884.1 | 3 | 1910 | |||
XM_008760148.1 | XP_008758370.1 | 2 | 2189 | |||
XM_008760149.1 | XP_008758371.1 | 2 | 1845 | |||
GenBank mRNA | BC083775.1 | - | 2 | 827 | 7193466 |