Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trub1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012173.1 | NP_001012173.1 | 6 | 613 | ||
XM_008760538.1 | XP_008758760.1 | 6 | 644 | |||
GenBank mRNA | BC087683.1 | - | 6 | 613 | 7320660 |