Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ostc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108566.1 | NP_001102036.1 | 1, 2 | 186 | ||
GenBank mRNA | BC159425.1 | - | 1, 2 | 172 | 9030828 | |
FQ219812.1 | - | 1, 2 | 192 | |||
FQ219920.1 | - | 1, 2 | 200 | |||
FQ220255.1 | - | 1, 2 | 168 | |||
FQ230162.1 | - | 1, 2 | 168 | |||
FQ234814.1 | - | 1, 2 | 148 |