Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PROM1 SIRNA Assays ›
Gene Symbol:
PROM1
Gene Name:
prominin 1
Gene Aliases:
AC133, CD133, CORD12, MCDR2, MSTP061, PROML1, RP41, STGD4
Chromosome Location:
Chr.4: 15968226 - 16084100 on Build GRCh38
UniGene ID:
Hs.614734
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s16881
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
1 nmol
1 nmol
5 nmol
InVivo | 5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
20 nmol
40 nmol
250 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
InVivo | 1 umol
InVivo | 1 umol
InVivo | 1 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PROM1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001145847.1
NP_001139319.1
19, 20
2622
NM_001145848.1
NP_001139320.1
19, 20
2338
NM_001145849.1
NP_001139321.1
19, 20
2342
NM_001145850.1
NP_001139322.1
19, 20
2342
NM_001145851.1
NP_001139323.1
18, 19
2315
NM_001145852.1
NP_001139324.1
18, 19
2315
NM_006017.2
NP_006008.1
19, 20
2342
XM_005248195.4
XP_005248252.1
19, 20
2438
XM_005248196.4
XP_005248253.1
19, 20
2438
XM_006713974.3
XP_006714037.1
18, 19
1943
XM_011513890.1
XP_011512192.1
20, 21
2424
XM_011513892.2
XP_011512194.1
20, 21
2575
XM_011513893.2
XP_011512195.1
20, 21
2649
XM_011513894.2
XP_011512196.1
20, 21
2734
XM_011513895.2
XP_011512197.1
21, 22
2850
XM_011513896.2
XP_011512198.1
20, 21
2479
XM_011513897.2
XP_011512199.1
20, 21
2666
XM_011513898.2
XP_011512200.2
20, 21
2424
XM_011513899.2
XP_011512201.2
19, 20
2438
XM_011513900.2
XP_011512202.1
20, 21
2424
XM_011513902.2
XP_011512204.1
20, 21
2424
XM_011513903.2
XP_011512205.1
19, 20
1973
XM_017008799.1
XP_016864288.1
19, 20
2468
XM_017008800.1
XP_016864289.1
20, 21
2424
XM_017008801.1
XP_016864290.1
20, 21
2424
XM_017008802.1
XP_016864291.1
20, 21
2516
XM_017008803.1
XP_016864292.1
19, 20
2413
XM_017008804.1
XP_016864293.1
19, 20
2483
XM_017008805.1
XP_016864294.1
19, 20
2356
GenBank mRNA
AF027208.1
-
19, 20
2167
AF117225.1
-
19, 20
2383
AF507034.1
-
18, 19
2140
AK027420.1
-
19, 20
2361
AK027422.1
-
19, 20
2383
AK075411.1
-
19, 20
2338
AK226033.1
-
19, 20
2361
AK314529.1
-
19, 20
2382
AY449689.1
-
18, 19
2103
AY449690.1
-
18, 19
2103
AY449691.1
-
18, 19
2103
AY449692.1
-
19, 20
2130
AY449693.1
-
19, 20
2130
BC012089.1
-
19, 20
2338
4644690
HQ628626.1
-
18, 19
2103
HQ628627.1
-
20, 21
2359
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
retina layer formation
photoreceptor cell maintenance
retina morphogenesis in camera-type eye
camera-type eye photoreceptor cell differentiation
glomerular visceral epithelial cell differentiation
glomerular parietal epithelial cell differentiation
positive regulation of nephron tubule epithelial cell differentiation
protein binding
actinin binding
cadherin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0