Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KAT2B SIRNA Assays ›
          Gene Symbol:
          KAT2B
          Gene Name:
          lysine acetyltransferase 2B
          Gene Aliases:
          CAF, P/CAF, PCAF
          Chromosome Location:
          Chr.3: 20040032 - 20154404 on Build GRCh38
          UniGene ID:
          Hs.533055
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s16895
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 562.00
          Your Price
          Online offer: 533.65
          533.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KAT2B
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_003884.4 NP_003875.3 18 2793
          XM_005265528.4 XP_005265585.1 17 2333
          XM_017007423.1 XP_016862912.1 19 2248
          XM_017007424.1 XP_016862913.1 18 2344
          GenBank mRNA AL832173.1 - 8 706
          BC060823.1 - 18 2398 30336561
          BC070075.1 - 18 2463 30333414
          U57317.2 - 18 2805

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00187332_m1

          Gene Ontology Categories:

          Function(s) Process(es)

          chromatin remodeling
          transcription initiation from RNA polymerase II promoter
          protein acetylation
          cell cycle arrest
          Notch signaling pathway
          memory
          negative regulation of cell proliferation
          regulation of protein ADP-ribosylation
          positive regulation of neuron projection development
          viral process
          N-terminal peptidyl-lysine acetylation
          internal peptidyl-lysine acetylation
          peptidyl-lysine acetylation
          cellular response to insulin stimulus
          cellular response to oxidative stress
          positive regulation of chromatin binding
          positive regulation of gluconeogenesis by positive regulation of transcription from RNA polymerase II promoter
          histone H3 acetylation
          histone H3-K9 acetylation
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of gene expression, epigenetic
          positive regulation of vasodilation
          positive regulation of transcription from RNA polymerase II promoter
          rhythmic process
          cellular response to parathyroid hormone stimulus
          positive regulation of histone H3-K14 acetylation
          positive regulation of histone H3-K9 acetylation
          RNA polymerase II regulatory region sequence-specific DNA binding
          chromatin binding
          transcription cofactor activity
          transcription coactivator activity
          histone acetyltransferase activity
          lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
          cyclin-dependent protein serine/threonine kinase inhibitor activity
          protein binding
          transcription factor binding
          acetyltransferase activity
          protein kinase binding
          protein complex binding
          histone acetyltransferase binding
          histone deacetylase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0