Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Herc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108631.1 | NP_001102101.1 | 20 | 2903 | ||
XM_006236584.2 | XP_006236646.1 | 19 | 2799 | |||
XM_006236585.2 | XP_006236647.1 | 19 | 2799 |