Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ALDH1A2 SIRNA Assays ›
          Gene Symbol:
          ALDH1A2
          Gene Name:
          aldehyde dehydrogenase 1 family member A2
          Gene Aliases:
          RALDH(II), RALDH2, RALDH2-T
          Chromosome Location:
          Chr.15: 57953424 - 58065923 on Build GRCh38
          UniGene ID:
          Hs.643455
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s16907
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ALDH1A2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001206897.1 NP_001193826.1 4, 5 728
          NM_003888.3 NP_003879.2 3, 4 631
          NM_170696.2 NP_733797.1 3, 4 631
          NM_170697.2 NP_733798.1 1, 2 235
          GenBank mRNA AB015226.1 - 3, 4 401
          AB015227.1 - 1, 2 164
          AB015228.1 - 1, 2 126
          AK128709.1 - 3, 4 1126
          AK294981.1 - 3, 4 331
          AK303057.1 - 4, 5 473
          BC030589.2 - 3, 4 389 4826743
          BX443600.2 - 1, 2 235

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00180254_m1

          Panther Classification:

          Molecular Function -

          dehydrogenase oxidoreductase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          blood vessel development
          kidney development
          liver development
          regulation of endothelial cell proliferation
          retinoic acid biosynthetic process
          heart morphogenesis
          vitamin A metabolic process
          midgut development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          determination of bilateral symmetry
          anterior/posterior pattern specification
          proximal/distal pattern formation
          positive regulation of gene expression
          neural crest cell development
          morphogenesis of embryonic epithelium
          neural tube development
          pituitary gland development
          neuron differentiation
          lung development
          hindbrain development
          pancreas development
          embryonic camera-type eye development
          response to estradiol
          response to vitamin A
          response to cytokine
          embryonic forelimb morphogenesis
          ureter maturation
          retinol metabolic process
          retinoic acid metabolic process
          retinal metabolic process
          9-cis-retinoic acid biosynthetic process
          positive regulation of apoptotic process
          retinoic acid receptor signaling pathway
          embryonic digestive tract development
          cardiac muscle tissue development
          oxidation-reduction process
          face development
          cellular response to retinoic acid
          retinal dehydrogenase activity
          3-chloroallyl aldehyde dehydrogenase activity
          aldehyde dehydrogenase (NAD) activity
          retinal binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lq2bv:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0