Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017413.4 | NP_059109.3 | 3 | 974 | ||
GenBank mRNA | AK001855.1 | - | 2 | 541 | ||
BC021104.1 | - | 3 | 955 | 4586949 |