siRNA Details
Gene Symbol: Mup4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198784.1 | NP_942079.1 | 166 | |||
GenBank mRNA | AB039829.1 | - | 166 |

Gene Symbol: Mup4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198784.1 | NP_942079.1 | 166 | |||
GenBank mRNA | AB039829.1 | - | 166 |