Search
Search

Brd4| Gene Symbol: Brd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100903.1 | NP_001094373.1 | 2, 3 | 438 | ||
| XM_006241072.2 | XP_006241134.1 | 2, 3 | 442 | |||
| XM_006241073.2 | XP_006241135.1 | 2, 3 | 646 | |||
| XM_006241074.2 | XP_006241136.1 | 2, 3 | 1370 | |||
| XM_008765170.1 | XP_008763392.1 | 1, 2 | 2161 | |||
| XM_008765171.1 | XP_008763393.1 | 1, 2 | 2161 | |||
| XM_008765172.1 | XP_008763394.1 | 1, 2 | 2161 | |||
| GenBank mRNA | BC100641.1 | - | 2, 3 | 385 | ||