Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other YWHAE SIRNA Assays ›
          Gene Symbol:
          YWHAE YWHAEP1
          Gene Name:
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon
          tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein epsilon pseudogene 1
          Gene Aliases:
          14-3-3E, HEL2, KCIP-1, MDCR, MDS
          -
          Chromosome Location:
          Chr.17: 1344490 - 1400375 on Build GRCh38
          Chromosome Location:
          Chr.7: 64433693 - 64435520 on Build GRCh38
          UniGene ID:
          Hs.513851
          Hs.728871
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID s17
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: YWHAE
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006761.4 NP_006752.1 2 386
          NR_024058.1 3 419
          XM_005256784.3 XP_005256841.1 2 499
          XM_017025005.1 XP_016880494.1 3 369
          GenBank mRNA AK128785.1 - 2 344
          AK295260.1 - 3 254
          AK295997.1 - 2 331
          AK296050.1 - 2 330
          AK316185.1 - 3 377
          AY883089.1 - 3 346
          BC000179.2 - 2 266 2900956
          BC001440.2 - 2 312 3139004
          BM755263.1 - 2 349
          BT007161.1 - 2 234
          DC365395.1 - 2 387
          EU794581.1 - 2 312
          U20972.1 - 2 318
          U28936.1 - 2 234
          U43399.1 - 2 293
          U43430.1 - 2 293
          U54778.1 - 2 294
          Gene Symbol: YWHAEP1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_029404.1 1 406
          GenBank mRNA DB053774.1 - 1 365

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00356749_g1

          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          neuron migration
          regulation of heart rate by hormone
          protein targeting
          viral process
          substantia nigra development
          hippocampus development
          cerebral cortex development
          hippo signaling
          intracellular signal transduction
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of membrane repolarization
          membrane organization
          membrane repolarization during cardiac muscle cell action potential
          regulation of heart rate by cardiac conduction
          cell-cell adhesion
          regulation of cellular response to heat
          positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
          regulation of potassium ion transmembrane transporter activity
          negative regulation of peptidyl-serine dephosphorylation
          protein binding
          potassium channel regulator activity
          enzyme binding
          protein domain specific binding
          MHC class II protein complex binding
          ubiquitin protein ligase binding
          histone deacetylase binding
          ion channel binding
          poly(A) RNA binding
          protein heterodimerization activity
          phosphoserine binding
          phosphoprotein binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0