Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ripply2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006243504.2 | XP_006243566.1 | 3 | 1527 | ||
XM_006243505.2 | XP_006243567.1 | 4 | 407 | |||
XM_008766459.1 | XP_008764681.1 | 4 | 1577 | |||
XM_343444.6 | XP_343445.2 | 4 | 1290 |