Search
Search

Pcbp4| Gene Symbol: Pcbp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191883.1 | NP_001178812.1 | 5 | 452 | ||
| XM_006243802.2 | XP_006243864.1 | 6 | 948 | |||
| XM_006243803.2 | XP_006243865.1 | 6 | 947 | |||
| GenBank mRNA | AW533143.1 | - | 5 | 382 | ||