Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prrg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191888.1 | NP_001178817.1 | 5 | 491 | ||
XM_006256963.2 | XP_006257025.1 | 6 | 655 | |||
XM_006256965.2 | XP_006257027.1 | 4 | 485 |