Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St6galnac5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271361.1 | NP_001258290.1 | 987 | |||
GenBank mRNA | AJ646872.1 | - | 757 |