Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PIAS2 SIRNA Assays ›
Gene Symbol:
PIAS2
Gene Name:
protein inhibitor of activated STAT 2
Gene Aliases:
ARIP3, DIP, MIZ, MIZ1, PIASX, PIASX-ALPHA, PIASX-BETA, SIZ2, ZMIZ4
Chromosome Location:
Chr.18: 46803224 - 46920167 on Build GRCh38
UniGene ID:
Hs.57769
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17285
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
5,366.00
Your Price
Online offer:
5366.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PIAS2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001324046.1
NP_001310975.1
9
1268
NM_001324047.1
NP_001310976.1
8
1179
NM_001324048.1
NP_001310977.1
9
1268
NM_001324049.1
NP_001310978.1
10
1391
NM_001324051.1
NP_001310980.1
8
1179
NM_001324052.1
NP_001310981.1
10
1369
NM_001324053.1
NP_001310982.1
10
1528
NM_001324054.1
NP_001310983.1
8
1159
NM_001324055.1
NP_001310984.1
9
1287
NM_001324057.1
NP_001310986.1
10
1369
NM_001324058.1
NP_001310987.1
7
1070
NM_001324059.1
NP_001310988.1
9
1268
NM_001324060.1
NP_001310989.1
7
968
NM_004671.4
NP_004662.2
7
1070
NM_173206.3
NP_775298.1
7
1070
NR_136684.1
6
985
NR_136685.1
6
595
XM_005258377.3
XP_005258434.1
7
968
XM_005258379.3
XP_005258436.1
7
1085
XM_006722571.3
XP_006722634.1
8
1053
XM_006722572.3
XP_006722635.1
8
1053
XM_006722573.2
XP_006722636.1
8
1053
XM_011526258.2
XP_011524560.1
7
1133
XM_011526259.2
XP_011524561.1
8
984
XM_017026068.1
XP_016881557.1
10
1369
XM_017026069.1
XP_016881558.1
8
1102
XM_017026070.1
XP_016881559.1
10
1369
XM_017026071.1
XP_016881560.1
8
984
XM_017026072.1
XP_016881561.1
7
923
GenBank mRNA
AF077953.1
-
7
883
AF077954.1
-
7
883
AF361054.1
-
7
961
AK294801.1
-
6
568
AK308143.1
-
6
907
BC015190.1
-
7
1041
4080297
BC111060.1
-
7
1012
5745101
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
protein sumoylation
androgen receptor signaling pathway
regulation of osteoblast differentiation
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
negative regulation of androgen receptor signaling pathway
DNA binding
transcription coactivator activity
protein binding
transcription factor binding
zinc ion binding
ligase activity
SUMO transferase activity
ubiquitin protein ligase binding
androgen receptor binding
SUMO ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0