Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LDB2 SIRNA Assays ›
          Gene Symbol:
          LDB2
          Gene Name:
          LIM domain binding 2
          Gene Aliases:
          CLIM1, LDB-2, LDB1
          Chromosome Location:
          Chr.4: 16501534 - 16898809 on Build GRCh38
          UniGene ID:
          Hs.714330
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s17323

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LDB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130834.2 NP_001124306.1 1 424
          NM_001290.4 NP_001281.1 1 424
          NM_001304434.1 NP_001291363.1 1 424
          NM_001304435.1 NP_001291364.1 1 424
          NR_130734.1 1 424
          XM_005248197.3 XP_005248254.1 1 394
          XM_005248201.2 XP_005248258.1 1 394
          XM_005248202.2 XP_005248259.1 1 394
          XM_006713975.3 XP_006714038.1 1 339
          XM_006713976.3 XP_006714039.1 1 340
          XM_006713977.3 XP_006714040.1 1 339
          XM_006713978.3 XP_006714041.1 1 340
          XM_006713979.3 XP_006714042.1 1 340
          XM_006713980.3 XP_006714043.1 1 341
          XM_006713981.3 XP_006714044.1 1 340
          XM_006713982.3 XP_006714045.1 1 340
          XM_006713983.3 XP_006714046.1 1 340
          XM_011513905.2 XP_011512207.1 1 293
          XM_017008812.1 XP_016864301.1 1 340
          XM_017008813.1 XP_016864302.1 1 394
          XM_017008814.1 XP_016864303.1 1 341
          XM_017008815.1 XP_016864304.1 1 341
          XM_017008816.1 XP_016864305.1 1 341
          XM_017008817.1 XP_016864306.1 1 394
          XM_017008819.1 XP_016864308.1 1 394
          XM_017008820.1 XP_016864309.1 1 394
          XM_017008821.1 XP_016864310.1 1 394
          XM_017008822.1 XP_016864311.1 1 249
          GenBank mRNA AF047337.1 - 1 209
          AF052389.1 - 2 282
          AF064492.1 - 1 209
          AF064493.1 - 1 209
          AF068651.1 - 1 207
          AK226029.1 - 1 249
          AK300119.1 - 1 21
          AK312881.1 - 1 119
          BC034019.1 - 1 217 4693654
          BG704168.1 - 1 324
          DA166898.1 - 1 416
          DA521198.1 - 1 424

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001106009, XM_008770201, XM_008770199, XM_008770202, XM_008770203, and others.
          This siRNA also targets the following transcripts in Mouse: NM_010698, NM_001077398, NM_001286348, AK146860, BC079611, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          hair follicle development
          epithelial structure maintenance
          regulation of cell migration
          somatic stem cell population maintenance
          regulation of kinase activity
          positive regulation of cellular component biogenesis
          positive regulation of transcription from RNA polymerase II promoter
          transcription cofactor activity
          enzyme binding
          LIM domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-s9xdw:80/100.66.76.163:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0