Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMSB4Y | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004202.2 | NP_004193.1 | 1 | 859 | ||
GenBank mRNA | AF000989.1 | - | 1 | 857 | ||
BC022482.1 | - | 1 | 122 | |||
BC119763.1 | - | 1 | 617 | 40119344 | ||
BC119764.1 | - | 1 | 617 | 40119345 |