Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103694517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773575.1 | XP_008771797.1 | 1 | 368 |
Gene Symbol: LOC100364877 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752159.3 | XP_003752207.1 | 1 | 225 |
Gene Symbol: LOC103690904 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773582.1 | XP_008771804.1 | 1 | 225 |
Gene Symbol: LOC103694516 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773552.1 | XP_008771774.1 | 1 | 225 |
Gene Symbol: LOC103694587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773722.1 | XP_008771944.1 | 1 | 225 |
Gene Symbol: LOC681067 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257516.2 | XP_006257578.1 | 1 | 434 |
Gene Symbol: LOC688702 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257588.2 | XP_006257650.1 | 1 | 416 |
Gene Symbol: LOC691030 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008773536.1 | XP_008771758.1 | 1 | 363 |
Gene Symbol: RGD1561558 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_228669.5 | XP_228669.3 | 225 |
Gene Symbol: RGD1565430 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006257538.2 | XP_006257600.1 | 370 | |||
XM_008773538.1 | XP_008771760.1 | 370 |