Search
Search

LATS1| Gene Symbol: LATS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004690.3 | NP_004681.1 | 8 | 3699 | ||
| NR_073033.1 | 9 | 3605 | ||||
| XM_017011474.1 | XP_016866963.1 | 9 | 3640 | |||
| XM_017011475.1 | XP_016866964.1 | 9 | 3560 | |||
| XM_017011476.1 | XP_016866965.1 | 10 | 3708 | |||
| XM_017011478.1 | XP_016866967.1 | 9 | 3618 | |||
| XM_017011479.1 | XP_016866968.1 | 9 | 3665 | |||
| XM_017011480.1 | XP_016866969.1 | 8 | 3337 | |||
| XM_017011481.1 | XP_016866970.1 | 7 | 3383 | |||
| GenBank mRNA | AF104413.1 | - | 8 | 3699 | ||
| AF164041.1 | - | 7 | 3183 | |||
| AK310422.1 | - | 9 | 3605 | |||