Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004707.3 | NP_004698.3 | 2 | 527 | ||
NR_033362.1 | 3 | 644 | ||||
NR_033363.1 | 3 | 643 | ||||
NR_073603.1 | 4 | 760 | ||||
GenBank mRNA | AB017507.1 | - | 2 | 247 | ||
AK098251.1 | - | 2 | 1697 | |||
AK313320.1 | - | 2 | 522 | |||
BC012266.2 | - | 2 | 478 | 3864058 | ||
BP335591.1 | - | 2 | 526 | |||
CB995198.1 | - | 3 | 355 | |||
CN400030.1 | - | 3 | 278 | |||
CR978239.1 | - | 4 | 453 | |||
DA005347.1 | - | 2 | 523 | |||
EU554057.1 | - | 2 | 369 |