Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vom1r78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008924.1 | NP_001008924.1 | 1 | 865 | ||
GenBank mRNA | AY510292.1 | - | 1 | 865 |
Gene Symbol: Vom1r77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008926.1 | NP_001008926.1 | 1 | 865 | ||
GenBank mRNA | AY510297.1 | - | 1 | 865 |