Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004218.3 | NP_004209.2 | 2 | 202 | ||
GenBank mRNA | AF498947.1 | - | 2 | 106 | ||
AK225036.1 | - | 2 | 120 | |||
AK225642.1 | - | 2 | 122 | |||
AK297498.1 | - | 2 | 130 | |||
AK312994.1 | - | 2 | 156 | |||
BC110081.1 | - | 2 | 108 | 40035112 | ||
BP367041.1 | - | 2 | 202 | |||
BT019535.1 | - | 2 | 106 | |||
BT019536.1 | - | 2 | 106 | |||
BX647356.1 | - | 2204 | ||||
CR536494.1 | - | 2 | 106 | |||
CR541691.1 | - | 2 | 106 | |||
D38516.1 | - | 2 | 115 | |||
EF560724.1 | - | 2 | 185 | |||
X79780.1 | - | 2 | 112 |