siRNA Details
Gene Symbol: LOC498465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251668.2 | XP_006251730.1 | 5 | 1468 | ||
XM_006251670.2 | XP_006251732.1 | 6 | 1343 | |||
GenBank mRNA | BC079195.1 | - | 5 | 1169 |
Gene Symbol: LOC102547093 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251682.2 | XP_006251744.1 | 5 | 1120 |
