Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMBL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289979.1 | NP_001276908.1 | 5 | 563 | ||
NM_001289980.1 | NP_001276909.1 | 5 | 447 | |||
NM_004756.4 | NP_004747.1 | 6 | 648 | |||
GenBank mRNA | AF015041.1 | - | 6 | 682 | ||
AK289398.1 | - | 6 | 640 | |||
BC001794.1 | - | 5 | 536 | |||
BC144583.1 | - | 5 | 458 | |||
BT009807.1 | - | 4 | 350 |