siRNA Details
Gene Symbol: LOC498465 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC079195.1 | - | 5 | 1063 |
Gene Symbol: LOC102546376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770586.1 | XP_008768808.1 | 5 | 1297 |
Gene Symbol: LOC102547212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770589.1 | XP_008768811.1 | 4 | 707 |
Gene Symbol: LOC102550466 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770593.1 | XP_008768815.1 | 5 | 1202 |
