Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SRSF11 SIRNA Assays ›
Gene Symbol:
SRSF11
Gene Name:
serine and arginine rich splicing factor 11
Gene Aliases:
NET2, SFRS11, dJ677H15.2, p54
Chromosome Location:
Chr.1: 70205682 - 70252020 on Build GRCh38
UniGene ID:
Hs.479693
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17770
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SRSF11
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001190987.1
NP_001177916.1
7
794
NM_004768.3
NP_004759.1
7
797
XM_005271338.3
XP_005271395.1
7
788
XM_005271339.3
XP_005271396.1
7
790
XM_011542427.2
XP_011540729.1
6
950
XM_011542428.2
XP_011540730.1
6
4186
XM_011542431.2
XP_011540733.1
7
1950
XM_017002811.1
XP_016858300.1
7
788
XM_017002812.1
XP_016858301.1
7
788
XM_017002813.1
XP_016858302.1
7
790
XM_017002814.1
XP_016858303.1
5
1919
XM_017002815.1
XP_016858304.1
7
3531
XM_017002816.1
XP_016858305.1
8
3665
XM_017002817.1
XP_016858306.1
8
3239
XM_017002818.1
XP_016858307.1
7
1067
XM_017002819.1
XP_016858308.1
8
1176
XM_017002820.1
XP_016858309.1
7
3894
XM_017002821.1
XP_016858310.1
7
1067
XM_017002822.1
XP_016858311.1
5
2282
XM_017002823.1
XP_016858312.1
6
1627
XM_017002824.1
XP_016858313.1
8
3665
XM_017002825.1
XP_016858314.1
8
3239
XM_017002826.1
XP_016858315.1
7
1067
XM_017002827.1
XP_016858316.1
8
1176
XM_017002828.1
XP_016858317.1
9
3010
XM_017002829.1
XP_016858318.1
9
3046
XM_017002830.1
XP_016858319.1
7
4171
XM_017002831.1
XP_016858320.1
8
3239
XM_017002832.1
XP_016858321.1
7
1067
XM_017002833.1
XP_016858322.1
8
1176
XM_017002834.1
XP_016858323.1
6
1909
GenBank mRNA
AK301670.1
-
7
965
AL832279.1
-
4
1056
BC002784.2
-
6
808
BC009840.2
-
6
791
BC017359.2
-
6
914
BC022815.1
-
7
794
BC032727.1
-
7
781
BC040436.1
-
7
752
5518580
BX640645.1
-
5
2034
BX641004.1
-
3
217
M74002.1
-
7
756
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
mRNA processing
RNA export from nucleus
mRNA export from nucleus
RNA splicing
mRNA 3'-end processing
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0