Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other KLF4 SIRNA Assays ›
          Gene Symbol:
          KLF4
          Gene Name:
          Kruppel like factor 4
          Gene Aliases:
          EZF, GKLF
          Chromosome Location:
          Chr.9: 107484852 - 107489720 on Build GRCh38
          UniGene ID:
          Hs.376206
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s17795

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price 5,366.00
          Your Price
          Online offer:
          5366.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: KLF4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001314052.1 NP_001300981.1 3 840
          NM_004235.5 NP_004226.3 3 840
          GenBank mRNA AF022184.1 - 3 691
          AF105036.1 - 3 594
          AK095134.1 - 1 346
          AK313489.1 - 3 429
          BC029923.1 - 3 405 5111134
          BC030811.1 - 2 294 4703002
          U70663.1 - 3 768

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C2H2 zinc finger transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          mesodermal cell fate determination
          negative regulation of cell proliferation
          epidermal cell differentiation
          positive regulation of gene expression
          negative regulation of gene expression
          negative regulation of phosphatidylinositol 3-kinase signaling
          negative regulation of muscle hyperplasia
          stem cell population maintenance
          post-embryonic camera-type eye development
          negative regulation of NF-kappaB transcription factor activity
          positive regulation of cellular protein metabolic process
          negative regulation of heterotypic cell-cell adhesion
          somatic stem cell population maintenance
          post-embryonic hemopoiesis
          negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
          regulation of phosphatidylinositol 3-kinase activity
          negative regulation of interleukin-8 biosynthetic process
          positive regulation of nitric oxide biosynthetic process
          fat cell differentiation
          regulation of cell differentiation
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of hemoglobin biosynthetic process
          negative regulation of smooth muscle cell proliferation
          regulation of axon regeneration
          epidermis morphogenesis
          negative regulation of inflammatory response
          positive regulation of protein metabolic process
          negative regulation of protein kinase B signaling
          positive regulation of telomerase activity
          canonical Wnt signaling pathway
          negative regulation of response to cytokine stimulus
          cellular response to hydrogen peroxide
          negative regulation of ERK1 and ERK2 cascade
          cellular response to retinoic acid
          cellular response to growth factor stimulus
          cellular response to cycloheximide
          cellular response to laminar fluid shear stress
          negative regulation of cell migration involved in sprouting angiogenesis
          cellular response to peptide
          negative regulation of leukocyte adhesion to arterial endothelial cell
          negative regulation of chemokine (C-X-C motif) ligand 2 production
          positive regulation of transcription regulatory region DNA binding
          core promoter proximal region sequence-specific DNA binding
          transcription factor activity, sequence-specific DNA binding transcription factor recruiting
          core promoter binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription factor binding
          transcriptional activator activity, RNA polymerase II transcription factor binding
          transcription cofactor binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          beta-catenin binding
          zinc ion binding
          phosphatidylinositol 3-kinase regulator activity
          histone deacetylase binding
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0