Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006233882.2 | XP_006233944.1 | 7 | 2082 | ||
XM_006233883.2 | XP_006233945.1 | 7 | 2082 |