Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CNOT8 SIRNA Assays ›
          Gene Symbol:
          CNOT8
          Gene Name:
          CCR4-NOT transcription complex subunit 8
          Gene Aliases:
          CAF1, CALIF, Caf1b, POP2, hCAF1
          Chromosome Location:
          Chr.5: 154857952 - 154876792 on Build GRCh38
          UniGene ID:
          Hs.26703
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s17849

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CNOT8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001301073.1 NP_001288002.1 2 413
          NM_001301074.1 NP_001288003.1 2 413
          NM_001301075.1 NP_001288004.1 2 461
          NM_004779.5 NP_004770.4 2 461
          XM_005268527.2 XP_005268584.1 3 915
          XM_011537706.1 XP_011536008.1 2 334
          XM_017010048.1 XP_016865537.1 3 320
          XM_017010049.1 XP_016865538.1 3 915
          XM_017010050.1 XP_016865539.1 3 900
          XM_017010051.1 XP_016865540.1 3 950
          XM_017010052.1 XP_016865541.1 3 867
          XM_017010053.1 XP_016865542.1 3 885
          XM_017010054.1 XP_016865543.1 2 406
          XM_017010055.1 XP_016865544.1 2 438
          XM_017010056.1 XP_016865545.1 2 524
          XM_017010057.1 XP_016865546.1 2 377
          GenBank mRNA AF087844.1 - 1 163
          AF180476.1 - 1 163
          AK025358.1 - 2 382
          AK297381.1 - 2 326
          AK299589.1 - 3 293
          AK303775.1 - 2 280
          AK314317.1 - 2 413
          AL122045.1 - 2 342
          AL520837.3 - 3 556
          BC017366.2 - 2 272 3506054
          BT006857.1 - 1 91

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          mRNA polyadenylation factor

          Gene Ontology Categories:

          Function(s) Process(es)

          nuclear-transcribed mRNA poly(A) tail shortening
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          gene silencing by miRNA
          exonucleolytic nuclear-transcribed mRNA catabolic process involved in deadenylation-dependent decay
          positive regulation of mRNA catabolic process
          RNA phosphodiester bond hydrolysis, exonucleolytic
          3'-5'-exoribonuclease activity
          transcription factor activity, sequence-specific DNA binding
          RNA binding
          poly(A)-specific ribonuclease activity
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-kmb4w:80/100.66.76.53:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0