Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109205.1 | NP_001102675.1 | 1 | 342 | ||
GenBank mRNA | BC162030.1 | - | 1 | 278 | 9100917 |