Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191967.1 | NP_001178896.1 | 2 | 136 | ||
XM_006235798.1 | XP_006235860.1 | 2 | 147 |