Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NRXN3 SIRNA Assays ›
Gene Symbol:
NRXN3
Gene Name:
neurexin 3
Gene Aliases:
C14orf60
Chromosome Location:
Chr.14: 77979904 - 79868290 on Build GRCh38
UniGene ID:
Hs.368307
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s17923
Purification
Standard
HPLC
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NRXN3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105250.2
NP_001098720.1
2
1236
NM_001272020.1
NP_001258949.1
2
1236
NM_004796.5
NP_004787.2
13
2689
NM_138970.4
NP_620426.2
2
1236
NR_073546.1
2
1236
NR_073547.1
15
4270
XM_005268218.2
XP_005268275.1
18
4339
XM_006720322.2
XP_006720385.1
17
4327
XM_006720323.2
XP_006720386.1
16
4303
XM_011537363.1
XP_011535665.1
18
4339
XM_011537364.2
XP_011535666.1
19
4301
XM_011537365.2
XP_011535667.1
18
4127
XM_011537366.1
XP_011535668.1
17
4315
XM_011537367.1
XP_011535669.1
18
4339
XM_011537368.1
XP_011535670.1
18
4339
XM_011537369.1
XP_011535671.1
18
4339
XM_011537370.1
XP_011535672.1
17
4315
XM_011537371.1
XP_011535673.1
18
4339
XM_011537372.1
XP_011535674.1
18
4339
XM_011537373.1
XP_011535675.1
18
4339
XM_011537377.2
XP_011535679.1
14
2631
XM_017021790.1
XP_016877279.1
19
4439
XM_017021791.1
XP_016877280.1
19
4222
XM_017021792.1
XP_016877281.1
17
4122
XM_017021793.1
XP_016877282.1
17
4312
XM_017021794.1
XP_016877283.1
16
4303
XM_017021795.1
XP_016877284.1
16
4098
XM_017021796.1
XP_016877285.1
16
4288
XM_017021797.1
XP_016877286.1
15
4279
XM_017021798.1
XP_016877287.1
15
4276
XM_017021799.1
XP_016877288.1
18
4339
XM_017021800.1
XP_016877289.1
18
4339
XM_017021801.1
XP_016877290.1
17
4312
XM_017021802.1
XP_016877291.1
16
4333
XM_017021803.1
XP_016877292.1
17
4318
XM_017021804.1
XP_016877293.1
17
4312
XM_017021805.1
XP_016877294.1
16
4303
XM_017021806.1
XP_016877295.1
16
4288
XM_017021807.1
XP_016877296.1
18
4339
GenBank mRNA
AB018286.1
-
13
2670
AJ316284.2
-
15
4058
AJ493127.1
-
2
312
AK056530.1
-
2
1236
AL833739.1
-
2
531
BC059368.1
-
2
1162
BC068469.1
-
2
1162
BC142649.1
-
2
669
BC150194.1
-
2
669
BC152457.1
-
13
2670
8862590
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neuron cell-cell adhesion
neurotransmitter secretion
axon guidance
synapse assembly
learning
adult behavior
social behavior
positive regulation of synapse assembly
vocalization behavior
positive regulation of synapse maturation
receptor activity
calcium channel regulator activity
metal ion binding
cell adhesion molecule binding
neuroligin family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0