Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esrp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127564.2 | NP_001121036.2 | 12 | 1790 | ||
XM_006237876.2 | XP_006237938.1 | 12 | 1870 | |||
XM_006237877.2 | XP_006237939.1 | 12 | 1870 | |||
XM_006237878.2 | XP_006237940.1 | 12 | 1870 | |||
XM_006237879.2 | XP_006237941.1 | 12 | 1869 | |||
XM_006237880.2 | XP_006237942.1 | 12 | 1869 | |||
GenBank mRNA | BC166735.1 | - | 12 | 1790 | 7936665 |