Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109265.1 | NP_001102735.1 | 3 | 1202 | ||
XM_006238967.1 | XP_006239029.1 | 4 | 1264 |