Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serhl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130579.1 | NP_001124051.1 | 7 | 554 | ||
XM_008765775.1 | XP_008763997.1 | 7 | 540 |