Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048243.2 | NP_001041708.2 | 3, 4 | 264 | ||
XM_006242661.2 | XP_006242723.1 | 3, 4 | 445 | |||
GenBank mRNA | BC157818.1 | - | 2, 3 | 167 | 7936655 | |
CB713235.1 | - | 3, 4 | 264 |