Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MAP4K4 SIRNA Assays ›
Gene Symbol:
MAP4K4
Gene Name:
mitogen-activated protein kinase kinase kinase kinase 4
Gene Aliases:
FLH21957, HEL-S-31, HGK, MEKKK4, NIK
Chromosome Location:
Chr.2: 101697703 - 101894690 on Build GRCh38
UniGene ID:
Hs.701013
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s18097
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MAP4K4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242559.1
NP_001229488.1
24
2903
NM_001242560.1
NP_001229489.1
25
3264
NM_004834.4
NP_004825.3
24
3030
NM_145686.3
NP_663719.2
25
3378
NM_145687.3
NP_663720.1
25
2822
XM_005264048.2
XP_005264105.1
27
3407
XM_005264050.3
XP_005264107.1
26
3245
XM_005264052.3
XP_005264109.1
26
3242
XM_005264057.3
XP_005264114.1
26
3176
XM_005264058.3
XP_005264115.1
25
3167
XM_005264059.3
XP_005264116.1
26
3083
XM_005264060.3
XP_005264117.1
25
3071
XM_005264062.3
XP_005264119.1
25
3014
XM_005264063.3
XP_005264120.1
25
3011
XM_005264064.3
XP_005264121.1
24
3005
XM_005264065.3
XP_005264122.1
25
2921
XM_005264066.3
XP_005264123.1
25
2918
XM_005264068.3
XP_005264125.1
24
2912
XM_005264069.2
XP_005264126.1
24
2882
XM_005264071.2
XP_005264128.1
23
2810
XM_006712865.2
XP_006712928.1
26
3152
XM_006712868.2
XP_006712931.1
25
2921
XM_006712869.2
XP_006712932.1
24
2822
XM_017005349.1
XP_016860838.1
27
3311
XM_017005350.1
XP_016860839.1
26
3245
XM_017005351.1
XP_016860840.1
25
3164
XM_017005352.1
XP_016860841.1
26
3176
XM_017005353.1
XP_016860842.1
26
3173
XM_017005354.1
XP_016860843.1
26
3152
XM_017005355.1
XP_016860844.1
25
3143
XM_017005356.1
XP_016860845.1
25
3074
XM_017005357.1
XP_016860846.1
26
3083
XM_017005358.1
XP_016860847.1
26
3080
XM_017005359.1
XP_016860848.1
25
3053
XM_017005360.1
XP_016860849.1
25
3050
XM_017005361.1
XP_016860850.1
25
3053
XM_017005362.1
XP_016860851.1
25
3050
XM_017005363.1
XP_016860852.1
25
3014
XM_017005364.1
XP_016860853.1
24
3002
XM_017005365.1
XP_016860854.1
25
2984
XM_017005366.1
XP_016860855.1
25
2960
XM_017005367.1
XP_016860856.1
25
2957
XM_017005368.1
XP_016860857.1
25
2888
XM_017005369.1
XP_016860858.1
24
2819
XM_017005370.1
XP_016860859.1
24
2819
XM_017005371.1
XP_016860860.1
23
2813
XM_017005372.1
XP_016860861.1
24
2729
XM_017005373.1
XP_016860862.1
23
2720
XM_017005374.1
XP_016860863.1
24
2729
XM_017005375.1
XP_016860864.1
23
2720
GenBank mRNA
AB013385.1
-
8
810
AB014587.1
-
23
2706
AF096300.1
-
24
2731
AK001272.1
-
8
965
AK025610.1
-
12
1579
AK074592.1
-
4
571
AK295610.1
-
24
3048
AK296396.1
-
25
2960
AL137755.1
-
17
2359
AY212247.1
-
24
2898
AY834276.1
-
20
2307
EU794634.1
-
25
2872
Back To Top
More Information
Panther Classification:
Molecular Function -
non-receptor serine/threonine protein kinase
Gene Ontology Categories:
Function(s)
Process(es)
protein phosphorylation
signal transduction by protein phosphorylation
microvillus assembly
intracellular signal transduction
negative regulation of apoptotic process
regulation of JNK cascade
creatine kinase activity
protein serine/threonine kinase activity
receptor signaling protein serine/threonine kinase activity
protein binding
ATP binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-p89b7:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0