Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EIF2AK3 SIRNA Assays ›
          Gene Symbol:
          EIF2AK3
          Gene Name:
          eukaryotic translation initiation factor 2 alpha kinase 3
          Gene Aliases:
          PEK, PERK, WRS
          Chromosome Location:
          Chr.2: 88556740 - 88627576 on Build GRCh38
          UniGene ID:
          Hs.591589
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID s18102
          Validated

          Purification
          Size
          Availability Inventoried
          Catalog # 4390824
          Price 7,898.00
          Your Price
          Online offer:
          7898.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EIF2AK3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001313915.1 NP_001300844.1 4 561
          NM_004836.6 NP_004827.4 4 987
          XM_017005376.1 XP_016860865.1 5 879
          GenBank mRNA AF110146.1 - 4 754
          AF193339.1 - 4 984
          AK128659.1 - 5 740
          AK308312.1 - 3 355
          AK315287.1 - 4 892
          BC126354.1 - 4 781 8992070
          BC126356.1 - 4 778 8992072
          CN285475.1 - 3 314
          CR749382.1 - 4 562

          Back To Top

          More Information


          Matching Taqman Gene Expression Assay:


          Hs00178128_m1

          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          ossification
          angiogenesis
          chondrocyte development
          protein phosphorylation
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          ER overload response
          endoplasmic reticulum organization
          positive regulation of vascular endothelial growth factor production
          positive regulation of gene expression
          regulation of translational initiation by eIF2 alpha phosphorylation
          negative regulation of translation
          peptidyl-serine phosphorylation
          calcium-mediated signaling
          insulin secretion
          bone mineralization
          endoplasmic reticulum unfolded protein response
          endocrine pancreas development
          negative regulation of myelination
          negative regulation of translational initiation in response to stress
          cellular response to amino acid starvation
          response to endoplasmic reticulum stress
          eiF2alpha phosphorylation in response to endoplasmic reticulum stress
          PERK-mediated unfolded protein response
          cellular response to glucose starvation
          positive regulation of transcription from RNA polymerase I promoter
          protein autophosphorylation
          insulin-like growth factor receptor signaling pathway
          protein homooligomerization
          regulation of endoplasmic reticulum stress-induced eIF2 alpha phosphorylation
          cellular response to cold
          positive regulation of protein localization to nucleus
          regulation of endoplasmic reticulum stress-induced intrinsic apoptotic signaling pathway
          response to manganese-induced endoplasmic reticulum stress
          protein kinase activity
          protein serine/threonine kinase activity
          eukaryotic translation initiation factor 2alpha kinase activity
          protein binding
          ATP binding
          enzyme binding
          protein phosphatase binding
          identical protein binding
          protein homodimerization activity
          Hsp90 protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0