Search
Search

Atp5h| Gene Symbol: Atp5h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019383.2 | NP_062256.1 | 3, 4 | 275 | ||
| GenBank mRNA | BC059139.1 | - | 3, 4 | 279 | 6921557 | |
| BC078846.1 | - | 2, 3 | 227 | 7133871 | ||
| D10021.1 | - | 2, 3 | 247 | |||
| D13120.1 | - | 3, 4 | 268 | |||
| FQ211219.1 | - | 3, 4 | 286 | |||
| FQ216885.1 | - | 3, 4 | 275 | |||
| FQ217843.1 | - | 3, 4 | 286 | |||
| FQ222479.1 | - | 3, 4 | 268 | |||