Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037850.1 | NP_001032939.1 | 3 | 247 | ||
NM_001037852.1 | NP_001032941.1 | 3 | 307 | |||
GenBank mRNA | DQ012092.1 | - | 3 | 247 | ||
DQ012093.1 | - | 3 | 307 |
Gene Symbol: Spag11bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728419.4 | XP_002728465.2 | 3 | 434 | ||
XM_006253377.2 | XP_006253439.1 | 3 | 437 | |||
GenBank mRNA | AY600145.1 | - | 3 | 267 |